MilliporeSigma
  • The blockade of lipophagy pathway is necessary for docosahexaenoic acid to regulate lipid droplet turnover in hepatic stellate cells
The blockade of lipophagy pathway is necessary for docosahexaenoic acid to regulate lipid droplet turnover in hepatic stellate cells
Biomedicine and Pharmacotherapy (2019)
Qiu S, et al.