MilliporeSigma
  • Protein Expressions of the Small GTPase Rho Proteins in Pterygial Tissue and Leukocytes of Patients with Pterygium
Protein Expressions of the Small GTPase Rho Proteins in Pterygial Tissue and Leukocytes of Patients with Pterygium
Proceedings (2018)
Saracaloglu A, et al.