MilliporeSigma
  • The crystal structure of ribonuclease A in complex with thymidine-3?-monophosphate provides further insight into ligand binding
The crystal structure of ribonuclease A in complex with thymidine-3?-monophosphate provides further insight into ligand binding
Proteins: Structure, Function, and Bioinformatics (2010)
Doucet N, et al.