MilliporeSigma
  • Cellular bioenergetics changes in magnocellular neurons may affect copeptin expression in the late phase of sepsis
Cellular bioenergetics changes in magnocellular neurons may affect copeptin expression in the late phase of sepsis
Journal of Neuroimmunology (2014)
Oliveira-Pelegrin G R, et al.