MilliporeSigma
  • Increasing serum transferrin to reduce tissue iron overload due to ineffective erythropoiesis
Increasing serum transferrin to reduce tissue iron overload due to ineffective erythropoiesis
The Hematology Journal : The Official Journal of the European Haematology Association / EHA (2015)
Tolosano E