MilliporeSigma
  • Effect of minerals on activity of microbial uricase to reduce ammonia volatilization in poultry manure.
Effect of minerals on activity of microbial uricase to reduce ammonia volatilization in poultry manure.
Poultry Science (2003)
Kim WK and Patterson PH