MilliporeSigma
  • Effect of basic fibroblast growth factor on cardiomyocyte differentiation from mouse embryonic stem cells.
Effect of basic fibroblast growth factor on cardiomyocyte differentiation from mouse embryonic stem cells.
Saudi Medical Journal (2007)
Khezri S, et al.