MilliporeSigma
  • Thiamin fortification of bread-making flour: Retention in bread and levels in Australian commercial fortified bread varieties
Thiamin fortification of bread-making flour: Retention in bread and levels in Australian commercial fortified bread varieties
J. Food Compos. Anal. (2015)
Tiong S A, et al.