MilliporeSigma
  • Streaming of proteolytic enzyme solutions for wound debridement: a feasibility study
Streaming of proteolytic enzyme solutions for wound debridement: a feasibility study
Wounds : a compendium of clinical research and practice (2004)
Yaakobi T, et al.