MilliporeSigma
  • Colony organization in the green alga Botryococcus braunii (Race B) is specified by a complex extracellular matrix
Colony organization in the green alga Botryococcus braunii (Race B) is specified by a complex extracellular matrix
Eukaryotic Cell (2012)
Weiss TL, et al.