MilliporeSigma
  • Combined proteomics and transcriptomics identifies carboxypeptidase b1 and nuclear factor kappaB (NF-kappaB) associated proteins as putative biomarkers of metastasis in low grade breast cancer
Combined proteomics and transcriptomics identifies carboxypeptidase b1 and nuclear factor kappaB (NF-kappaB) associated proteins as putative biomarkers of metastasis in low grade breast cancer
Molecular and Cellular Proteomics (2015)
Bouchal P, et al.