MilliporeSigma
  • Mutations of cytochrome c identified in patients with thrombocytopenia THC4 affect both apoptosis and cellular bioenergetics
Mutations of cytochrome c identified in patients with thrombocytopenia THC4 affect both apoptosis and cellular bioenergetics
Biochimica et Biophysica Acta (BBA)-Molecular Basis of Disease (2014)
De Rocco D, et al.