MilliporeSigma
  • An evaluation of wet chemical methods for quantifying sulfur fractions in freshwater wetland peat 1
An evaluation of wet chemical methods for quantifying sulfur fractions in freshwater wetland peat 1
Limnology and Oceanography (1985)
Wieder RK, et al.