MilliporeSigma
  • Berberine-improved visceral white adipose tissue insulin resistance associated with altered sterol regulatory element-binding proteins, liver x receptors, and peroxisome proliferator-activated receptors transcriptional programs in diabetic hamsters.
Berberine-improved visceral white adipose tissue insulin resistance associated with altered sterol regulatory element-binding proteins, liver x receptors, and peroxisome proliferator-activated receptors transcriptional programs in diabetic hamsters.
Biological & pharmaceutical bulletin (2011-05-03)
Guo-Sheng Li, Xu-Han Liu, Hua Zhu, Lan Huang, Ya-Li Liu, Chun-Mei Ma, Chuan Qin
ABSTRACT
The diabetic "lipotoxicity" hypothesis presents that fat-induced visceral white adipose tissue insulin resistance plays a central role in the pathogenesis of type 2 diabetes. Berberine, a hypolipidemic agent, has been reported to have antidiabetic activities. The molecular mechanisms for this property are, however, not well clarified. Therefore in this study type 2 diabetic hamsters were induced by high-fat diet with low-dose streptozotocin. Then, we investigated the gene expression alterations and explored the molecular mechanisms underlying the therapeutic effect of berberine on fat-induced visceral white adipose tissue insulin resistance in diabetic hamsters by microarray analysis followed by real-time reverse transcription-polymerase chain reaction (RT-PCR) confirmation. Type 2 diabetic hamsters exhibited hyperglycemia and relative hyperinsulinemia, glucose intolerance, insulin resistance, intra-adipocyte lipid accumulation, significant increase in body weight and visceral white adipose tissue weight, abnormal serum adipokines levels, and deleterious dyslipidemia. Furthermore, they had increased sterol regulatory element-binding proteins (SREBPs) expression and decreased liver X receptors (LXRs) and peroxisome proliferator-activated receptors (PPARs) expression in visceral white adipose tissue. After 9-week berberine treatment, fat-induced insulin resistance and diabetic phenotype in type 2 diabetic hamsters were significantly improved. Compared with diabetic hamsters, expression of LXRs and PPARs significantly increased and SREBPs significantly decreased in visceral white adipose tissue from berberine-treated diabetic hamsters. These results suggest that altered visceral white adipose tissue LXRs, PPARs, and SREBPs transcriptional programs are involved in the therapeutic mechanisms of berberine on fat-induced visceral white adipose tissue insulin resistance in type 2 diabetic hamsters.
Sigma-Aldrich
Carboxymethylcellulose sodium salt, Medium viscosity
Millipore
Mouse Leptin ELISA, This Mouse Leptin ELISA is used to measure & quantify Leptin levels in Metabolism & Endocrine & Neuroscience research.