MilliporeSigma
  • Nicotinic cholinoceptor-mediated excitation in ambigual motoneurons of the rat.
Nicotinic cholinoceptor-mediated excitation in ambigual motoneurons of the rat.
Neuroscience (1991-01-01)
Y T Wang, R S Neuman, D Bieger
ABSTRACT
The purpose of this study was to determine if ambigual oesophageal motoneurons of the rat possess functional nicotinic cholinoceptors. In urethane anaesthetized rats, acetylcholine (20-50 pmol) delivered micropneumophoretically from multibarrelled pipettes to the compact formation of the nucleus ambiguus produced either synchronous or propulsive oesophageal contractions which were fully and reversibly blocked by dihydro-beta-erythroidine (8-10 pmol) but were resistant to D-tubocurarine and hexamethonium (10-20 pmol). 1,1-Dimethyl-4-phenyl-piperazinium but not muscarine (8 pmol) exerted an analogous agonist action. Ejection of glutamate at the same sites produced similar oesophageal responses which were, however, resistant to dihydro-beta-erythroidine. Acetylcholine applied 5-15 s prior to glutamate transiently facilitated the glutamate-evoked response. The facilitatory effect of acetylcholine was replicated by 1,1-dimethyl-4-phenyl-piperazinium but not muscarine and inhibited by dihydro-beta-erythroidine. Physostigmine, applied either intra-ambigually (10-20 pmol) or by intravenous injection (0.15-0.3 mumol/kg), enhanced both acetylcholine and glutamate-evoked responses. In brainstem transverse slices, application of acetylcholine and glutamate to quiescent ambigual neurons of the compact formation resulted in a rapid membrane depolarization associated with an increased membrane conductance and spiking. Under voltage clamp, both acetylcholine and glutamate elicited a net inward current. The depolarizing response of these neurons to acetylcholine was blocked by dihydro-beta-erythroidine (0.5-2 pmol), hexamethonium (0.2 mM) and D-tubocurarine (10 microM) and persisted in the presence of tetrodotoxin (10(-6) M) or Mn2+ (5 mM) in the bathing medium.(ABSTRACT TRUNCATED AT 250 WORDS)