MilliporeSigma
  • Intracerebroventricular administration of the beta3-adrenoceptor agonist CL 316243 causes Fos immunoreactivity in discrete regions of rat hypothalamus
Intracerebroventricular administration of the beta3-adrenoceptor agonist CL 316243 causes Fos immunoreactivity in discrete regions of rat hypothalamus
Neuroscience Letters (2000)
Castillo-Melendez M, et al.