MilliporeSigma
  • Extraction of tannins and saponins from plant sources and their effects on in vitro methanogenesis and rumen fermentation
Extraction of tannins and saponins from plant sources and their effects on in vitro methanogenesis and rumen fermentation
Journal of the Indonesian Tropical Animal Agriculture (2014)
Yuliana P, et al.