MilliporeSigma
  • Dominant negative insulin-like growth factor-1 receptor inhibits neointimal formation through suppression of vascular smooth muscle cell migration and proliferation, and induction of apoptosis.
Dominant negative insulin-like growth factor-1 receptor inhibits neointimal formation through suppression of vascular smooth muscle cell migration and proliferation, and induction of apoptosis.
Biochemical and biophysical research communications (2004-11-16)
Hyun-Joung Lim, Hyun-Young Park, Young-Guk Ko, Sea-Hyoung Lee, Seung-Yeon Cho, Eun Jig Lee, J Larry Jameson, Yangsoo Jang
ABSTRACT
Blocking of the IGF-1 signaling pathway targeting the IGF-1 receptor (IGF-1R) provides a potential treatment strategy for restenosis. In this study, we have examined the effects of a dominant negative IGF-1R (IGF-1Rt) on primary rat VSMCs in vitro and on injured rat carotid artery in vivo. Ad/IGF-1Rt infection inhibited VSMC migration and proliferation, and it also induced apoptosis by inhibiting phosphorylation of Akt and phosphorylation of ERK1/2. Consistent with the anti-proliferative and apoptotic effects in vitro, the Ad/IGF-1Rt infection markedly reduced neointimal formation in carotid injury model. Ad/IGF-1Rt treated carotid arteries exhibited a suppressed proliferation index, PCNA expression, and also were stained positive for TUNEL assay. These results indicate that a dominant negative IGF-1R has the potential to reduce neointimal formation of injured rats' carotid arteries. The delivery of dominant negative IGF-1R by adenoviral or other vectors may provide a useful strategy for inhibiting restenosis after angioplasty.
Sigma-Aldrich
ANTI-FLAG® M2-FITC antibody, Mouse monoclonal, clone M2, purified immunoglobulin, buffered aqueous solution