MilliporeSigma
  • Casein, Dephosphorylated, has been used in studies of modulators of circadian clocks. It has been shown that several molecules can inhibit casein kinase which results in the lenghthening of these clocks.
Casein, Dephosphorylated, has been used in studies of modulators of circadian clocks. It has been shown that several molecules can inhibit casein kinase which results in the lenghthening of these clocks.
Proceedings of the National Academy of Sciences of the USA (2011)
Chen, Z., et al.