MilliporeSigma
  • Repair of a chondral defect using a cell free scaffold in a young patient--a case report of successful scaffold transformation and colonisation.
Repair of a chondral defect using a cell free scaffold in a young patient--a case report of successful scaffold transformation and colonisation.
BMC Surgery
Schuettler, KF; Struewer, J; Rominger, MB; Rexin, P; Efe, T
Sigma-Aldrich
Anti-Collagen Type II Antibody, clone 6B3, clone 6B3, Chemicon®, from mouse