MilliporeSigma
  • Electron attachment to hydrated oligonucleotide dimers: guanylyl-3′,5′-cytidine and cytidylyl-3′,5′-guanosine.
Electron attachment to hydrated oligonucleotide dimers: guanylyl-3′,5′-cytidine and cytidylyl-3′,5′-guanosine.
Chemistry (Easton) (2010)
Gu J, Xie Y, Schaefer HF 3rd.