MilliporeSigma
  • Phosphatidylcholine protects against steatosis in mice but not non-alcoholic steatohepatitis.
Phosphatidylcholine protects against steatosis in mice but not non-alcoholic steatohepatitis.
Biochimica et biophysica acta (2011-07-13)
Lorissa J Niebergall, René L Jacobs, Todd Chaba, Dennis E Vance
ABSTRACT
Several studies suggest that low levels of hepatic phosphatidylcholine (PC) play a role in the pathogenesis of non-alcoholic steatohepatitis (NASH). CTP: phosphocholine cytidylyltransferase (CT) is the key regulatory enzyme in the CDP-choline pathway for PC biosynthesis. Liver-specific elimination of CTα (LCTα(-/-)) in mice fed a chow diet decreases very-low-density lipoprotein secretion, reduces lipid efflux from liver, and causes mild steatosis. We fed LCTα(-/-) mice a high fat diet to determine if impaired PC biosynthesis played a role in development of NASH. LCTα(-/-) mice developed NASH within one week of high fat feeding. Hepatic CTα deficiency caused hepatic steatosis, a 2-fold increase in ceramide mass, and a 20% reduction in PC content. In an attempt to prevent NASH, LCTα(-/-) mice were either injected daily with CDP-choline or fed the high fat diet supplemented with betaine. In addition, LCTα(-/-) mice were injected with adenoviruses expressing CTα. CDP-choline injections and adenoviral expression of CTα increased hepatic PC, while dietary betaine supplementation normalized hepatic triacylglycerol but did not alter hepatic PC mass in LCTα(-/-) mice. Interestingly, none of the treatments normalized hepatic ceramide mass or fully prevented the development of NASH in LCTα(-/-) mice. These results show that normalizing the amount of hepatic PC is not sufficient to prevent NASH in LCTα(-/-) mice.
Sigma-Aldrich
Cytidine 5′-diphosphocholine sodium salt hydrate, ≥99.0% (HPLC)
Pricing and availability is not currently available.