MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
Identification of a novel CaMKK substrate.
Biochemical and biophysical research communications (2011-06-07)
Tomohito Fujimoto, Naoya Hatano, Naohito Nozaki, Saki Yurimoto, Ryoji Kobayashi, Hiroshi Tokumitsu
ABSTRACT
Ca(2+)/calmodulin-dependent protein kinase kinase (CaMKK) phosphorylates and activates specific downstream protein kinases including CaMKI, CaMKIV and 5'-AMP-activated protein kinase. In order to examine the variety of CaMKK-mediated signaling pathways, we searched for novel CaMKK substrate(s) using N(6)-(1-methylbutyl)-ATP and genetically engineered CaMKKα mutant, CaMKKα (Phe(230)Gly), that was capable of utilizing this ATP analogue as a phosphate donor. Incubation of rat brain extracts with recombinant CaMKKα (Phe(230)Gly), but not with wild-type kinase, in the presence of N(6)-(1-methylbutyl)-ATP and Ca(2+)/CaM, induced significant threonine phosphorylation of a 50kDa protein as well as CaMKI phosphorylation at Thr(177). The 50kDa CaMKK substrate was partially purified by using serial column chromatography, and was identified as Syndapin I by LC-MS/MS analysis. We confirmed that recombinant Syndapin I was phosphorylated by CaMKKα and β isoforms at Thr(355)in vitro. Phosphorylation of HA-Syndapin I at Thr(355) in transfected HeLa cells was significantly induced by co-expression of constitutively active mutants of CaMKK isoforms. This is the first report that CaMKK is capable of phosphorylating a non-kinase substrate suggesting the possibility of CaMKK-mediated novel Ca(2+)-signaling pathways that are independent of downstream protein kinases.
Sigma-Aldrich
Adenosine 5′-monophosphate monohydrate, from yeast, ≥97%
Sigma-Aldrich
Adenosine 5′-monophosphate sodium salt, from yeast, ≥99%
Sigma-Aldrich
Adenosine 5′-monophosphate disodium salt, ≥99.0% (HPLC)