MilliporeSigma
  • Studies on selectin blockers. 7. Structure-activity relationships of sialyl Lewis X mimetics based on modified Ser-Glu dipeptides.
Studies on selectin blockers. 7. Structure-activity relationships of sialyl Lewis X mimetics based on modified Ser-Glu dipeptides.
Journal of medicinal chemistry (1998-10-23)
T Tsukida, H Moriyama, K Kurokawa, T Achiha, Y Inoue, H Kondo
ABSTRACT
We have previously found that heterochiral fucodipeptides, L-Ser-D-Glu (3a) and D-Ser-L-Glu (3b), exhibited up to 20-100 times more potency than a sialyl Lewis X (sLeX, 1) and a 3'-sulfated Lewis X analogue (2) toward E-selectin binding and have also proposed, from molecular dynamics calculation, that their strong activities would depend on a possible formation of the type II and/or type II' beta-turn of compounds 3a,b (Tsukida, T.; Hiramatsu, Y.; Tsujishita, H.; Kiyoi, T.; Yoshida, M.; Kurokawa, K.; Moriyama, H.; Ohmoto, H.; Wada, Y.; Saito, T.; Kondo, H. J. Med. Chem. 1997, 40, 3534-3541). To clarify our hypothesis, we synthesized several analogues of compounds 3a,b and investigated their structure-activity relationships. As a result, it was indicated that the type II and/or type II' beta-turn conformation would be a comparatively tight form and would play important roles in favorable binding to E-selectin. These findings indicate that sLeX mimetics with type II and type II' beta-turn dipeptides could be a useful methodology for the design of an active selectin blocker.