MilliporeSigma
  • Cloning, expression pattern and mapping to 12p 13.2 --> p13.1 of CLAPS3, a gene encoding a novel clathrin-adaptor small chain.
Cloning, expression pattern and mapping to 12p 13.2 --> p13.1 of CLAPS3, a gene encoding a novel clathrin-adaptor small chain.
Cytogenetics and cell genetics (1996-01-01)
T K Watanabe, F Shimizu, M Nagata, A Takaichi, T Fujiwara, Y Nakamura, E Takahashi, Y Hirai
ABSTRACT
From a human fetal-brain cDNA library we isolated a novel gene encoding a peptide homologous to clathrin-adaptor small chains in rat, mouse, and yeast. The cDNA, designated CLAPS3 (clathrin-associated/assembly/adaptor protein, small 3, 22 kDa), contained an open reading frame of 579 nucleotides encoding 193 amino acids. Northern-blot analysis revealed expression of a 1.35-kb transcript in all human tissues examined. This gene was mapped to chromosome bands 12p13.2 --> p13.1 by FISH.