MilliporeSigma
  • Different sympathetic pathways control the metabolism of distinct bone envelopes.
Different sympathetic pathways control the metabolism of distinct bone envelopes.
Bone
Caroline Bataille,Cedric Mauprivez,Eric Hay,Brigitte Baroukh,Adrian Brun et al.