MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • HSF-1 regulators DDL-1/2 link insulin-like signaling to heat-shock responses and modulation of longevity.
HSF-1 regulators DDL-1/2 link insulin-like signaling to heat-shock responses and modulation of longevity.
Cell (2012-01-24)
Wei-Chung Chiang, Tsui-Ting Ching, Hee Chul Lee, Carol Mousigian, Ao-Lin Hsu
ABSTRACT
Extended longevity is often correlated with increased resistance against various stressors. Insulin/IGF-1-like signaling (IIS) is known to have a conserved role in aging and cellular mechanisms against stress. In C. elegans, genetic studies suggest that heat-shock transcription factor HSF-1 is required for IIS to modulate longevity. Here, we report that the activity of HSF-1 is regulated by IIS. This regulation occurs at an early step of HSF-1 activation via two HSF-1 regulators, DDL-1 and DDL-2. Inhibition of DDL-1/2 increases longevity and thermotolerance in an hsf-1-dependent manner. Furthermore, biochemical analyses suggest that DDL-1/2 negatively regulate HSF-1 activity by forming a protein complex with HSF-1. The formation of this complex (DHIC) is affected by the phosphorylation status of DDL-1. Both the formation of DHIC and the phosphorylation of DDL-1 are controlled by IIS. Our findings point to DDL-1/2 as a link between IIS and the HSF-1 pathway.
Product Description
Millipore
Monoclonal Anti-HA−Agarose antibody produced in mouse, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)