MilliporeSigma
  • Effect of dephytinization on bioavailability of iron, calcium and zinc from infant cereals assessed in the Caco-2 cell model.
Effect of dephytinization on bioavailability of iron, calcium and zinc from infant cereals assessed in the Caco-2 cell model.
World journal of gastroenterology (2009-04-29)
Carmen Frontela, Maria-Laura Scarino, Simonetta Ferruzza, Gaspar Ros, Carmen Martínez
ABSTRACT
To test the effect of the dephytinization of three different commercial infant cereals on iron, calcium, and zinc bioavailability by estimating the uptake, retention, and transport by Caco-2 cells. Both dephytinized (by adding an exogenous phytase) and non-dephytinized infant cereals were digested using an in vitro digestion protocol adapted to the gastrointestinal conditions of infants younger than 6 mo. Mineral cell retention, transport, and uptake from infant cereals were measured using the soluble fraction of the simulated digestion and the Caco-2 cells. Dephytinization of infant cereals significantly increased (P < 0.05) the cell uptake efficiency (from 0.66%-6.05% to 3.93%-13%), retention (from 6.04%-16.68% to 14.75%-20.14%) and transport efficiency (from 0.14%-2.21% to 1.47%-6.02%), of iron, and the uptake efficiency (from 5.0%-35.4% to 7.3%-41.6%) and retention (from 4.05%-20.53% to 14.45%-61.3%) of zinc, whereas calcium only cell uptake showed a significant increase (P < 0.05) after removing phytate from most of the samples analyzed. A positive relationship (P < 0.05) between mineral solubility and the cell uptake and transport efficiencies was observed. Removing phytate from infant cereals had a beneficial effect on iron and zinc bioavailability when infant cereals were reconstituted with water. Since in developing countries cereal-based complementary foods for infants are usually consumed mixed with water, exogenous phytase additions could improve the nutritional value of this weaning food.
Sigma-Aldrich
Pancreatin from porcine pancreas, 4 × USP specifications