MilliporeSigma
  • Effects of ketamine and L-glutamic acid diethyl ester on spatial and nonspatial learning tasks in rats.
Effects of ketamine and L-glutamic acid diethyl ester on spatial and nonspatial learning tasks in rats.
Pharmacology, biochemistry, and behavior (1993-03-01)
R Lalonde, C C Joyal
ABSTRACT
An NMDA antagonist, ketamine, at the highest dose tested (15 mg/kg), impaired the acquisition of a hole-board spatial learning task but not the acquisition of a left-right alternation task. A non-NMDA (quisqualate) antagonist, L-glutamic acid diethyl ester (LGDE), did not impair the acquisition of either task. Both drugs had effects on different aspects of a go-no go discrimination task and a straight runway task, ketamine tending to activate and LGDE tending to slow rats. These results concur with previous research regarding the sensitivity of some spatial tasks to NMDA antagonism. Non-NMDA antagonists affect behavior without causing spatial deficits.
Sigma-Aldrich
L-Glutamic acid diethyl ester hydrochloride, 97%