MilliporeSigma
  • Novel route to stabilization of bioactive antioxidants by encapsulation in electrospun fibers of zein prolamine
Novel route to stabilization of bioactive antioxidants by encapsulation in electrospun fibers of zein prolamine
Food Hydrocolloids (2009)
Fernandez A, et al.