MilliporeSigma
  • Direct characterization of the maize starch synthase IIa product shows maltodextrin elongation occurs at the non-reducing end
Direct characterization of the maize starch synthase IIa product shows maltodextrin elongation occurs at the non-reducing end
Test (2016)
Larson ME, et al.