MilliporeSigma
  • Changes of AMPA receptor properties in the neocortex and hippocampus following pilocarpine-induced status epilepticus in rats
Changes of AMPA receptor properties in the neocortex and hippocampus following pilocarpine-induced status epilepticus in rats
Neuroscience (2016)
Malkin SL, et al.