MilliporeSigma
  • The selective complex behavior between soybean whey proteins and i-carrageenan and isolation of the major proteins of the soybean whey
The selective complex behavior between soybean whey proteins and i-carrageenan and isolation of the major proteins of the soybean whey
Food Hydrocolloids (2016)
Li X, et al.