MilliporeSigma
  • Hematopoietic-to-mesenchymal transition of adipose tissue macrophages is regulated by integrin beta and fabricated fibrin matrices
Hematopoietic-to-mesenchymal transition of adipose tissue macrophages is regulated by integrin beta and fabricated fibrin matrices
Adipocyte (2017)
Gavin KM, et al.