MilliporeSigma
  • Cell Transplantation as a Method to Investigate Spinal Cord Regeneration in Regenerative and Nonregenerative Xenopus Stages
Cell Transplantation as a Method to Investigate Spinal Cord Regeneration in Regenerative and Nonregenerative Xenopus Stages
Cold Spring Harbor Protocols (2018)
Mendez-Olivos EE and LarraIn J