MilliporeSigma
  • Expression and characterization of recombinant type 2 3alpha-hydroxysteroid dehydrogenase (HSD) from human prostate: demonstration of bifunctional 3alpha17beta-HSD activity and cellular distribution
Expression and characterization of recombinant type 2 3alpha-hydroxysteroid dehydrogenase (HSD) from human prostate: demonstration of bifunctional 3alpha17beta-HSD activity and cellular distribution
Molecular Endocrinology (1997)
Lin HK, et al.