MilliporeSigma
  • Bioluminescent assay for total bile acids in serum with use of bacterial luciferase.
Bioluminescent assay for total bile acids in serum with use of bacterial luciferase.
Clinical Chemistry (1983)
Styrelius I, et al.