MilliporeSigma
  • Endothelin receptor antagonists attenuate the inflammatory response of human pulmonary vascular smooth muscle cells to bacterial endotoxin
Endothelin receptor antagonists attenuate the inflammatory response of human pulmonary vascular smooth muscle cells to bacterial endotoxin
Journal of Pharmacology and Experimental Therapeutics (2013)
Knobloch J, et al.