MilliporeSigma
  • Induction of TNF-alpha release from human buffy coat cells by Pseudomonas aeruginosa is reduced by lung surfactant protein A.
Induction of TNF-alpha release from human buffy coat cells by Pseudomonas aeruginosa is reduced by lung surfactant protein A.
FEBS letters (1998-11-06)
T P Hickling, R B Sim, R Malhotra
ABSTRACT
Lipopolysaccharide (LPS) induction of TNF-alpha release is a central event in the pathophysiology of gram-negative bacterial septicaemia. Lung surfactant protein A (SP-A) mediates pathogen/host cell interactions. Binding of SP-A to Pseudomonas aeruginosa LPS and the effects of SP-A with LPS or whole bacteria on buffy coat cells were investigated. SP-A interacts with P. aeruginosa LPS in a concentration and calcium dependent manner, either through the lipid A portion of LPS or through another lectin/carbohydrate interaction. SP-A decreased TNF-alpha secretion induced by bacteria or LPS from buffy coat cells, in a concentration dependent manner.