MilliporeSigma
  • Cysteine in the manganous-isocitrate binding site of pig heart TPN-specific isocitrate dehydrogenase. I. Kinetics of chemical modification and properties of thiocyano enzyme.
Cysteine in the manganous-isocitrate binding site of pig heart TPN-specific isocitrate dehydrogenase. I. Kinetics of chemical modification and properties of thiocyano enzyme.
Archives of biochemistry and biophysics (1981-03-01)
R A Johanson, R F Colman