MilliporeSigma
  • Direct isolation of poly (A)+ RNA from 4 M guanidine thiocyanate-lysed cell extracts using locked nucleic acid-oligo (T) capture
Direct isolation of poly (A)+ RNA from 4 M guanidine thiocyanate-lysed cell extracts using locked nucleic acid-oligo (T) capture
Nucleic Acids Research (2004)
Jacobsen N, et al.