MilliporeSigma
  • Tethered ansa-bridged titanium complexes immobilized on 3-mercaptopropyl-functionalized silica gel and their application for the hydrosilylation of imines
Tethered ansa-bridged titanium complexes immobilized on 3-mercaptopropyl-functionalized silica gel and their application for the hydrosilylation of imines
Dalton Transactions (2012)
Gruber-Woelfler H, et al.