MilliporeSigma
  • Enzyme disintegration with spatial resolution reveals different distributions of sludge extracellular polymer substances
Enzyme disintegration with spatial resolution reveals different distributions of sludge extracellular polymer substances
Biotechnology for Biofuels (2016)
Lu Fan, et al.