MilliporeSigma
  • Necrostatin-1 reduces histopathology and improves functional outcome after controlled cortical impact in mice
Necrostatin-1 reduces histopathology and improves functional outcome after controlled cortical impact in mice
Journal of Cerebral Blood Flow and Metabolism (2008)
You Z, et al.