MilliporeSigma
  • Phospho-Delta-Np63alpha-responsive microRNAs contribute to the regulation of necroptosis in squamous cell carcinoma upon cisplatin exposure
Phospho-Delta-Np63alpha-responsive microRNAs contribute to the regulation of necroptosis in squamous cell carcinoma upon cisplatin exposure
Febs Letters (2015)
Ratovitski EA