MilliporeSigma
  • The complete amino acid sequence of bovine liver catalase and the partial sequence of bovine erythrocyte catalase
The complete amino acid sequence of bovine liver catalase and the partial sequence of bovine erythrocyte catalase
Archives of Biochemistry and Biophysics (1982)
Schroeder WA, et al.