MilliporeSigma
  • Type I collagen extracted from rat-tail and bovine Achilles tendon for dental application: a comparative study
Type I collagen extracted from rat-tail and bovine Achilles tendon for dental application: a comparative study
Asian Biomedicine : Research, Reviews and News (2011)
Techatanawat S, et al.