MilliporeSigma
  • Substrate specificity of acetylxylan esterase from Schizophyllum commune: mode of action on acetylated carbohydrates
Substrate specificity of acetylxylan esterase from Schizophyllum commune: mode of action on acetylated carbohydrates
Biochimica et Biophysica Acta (1996)
Biely P, et al.