MilliporeSigma
  • DNA condensation by poly-L-lysine at the single molecule level: role of DNA concentration and polymer length
DNA condensation by poly-L-lysine at the single molecule level: role of DNA concentration and polymer length
Journal of Controlled Release : Official Journal of the Controlled Release Society (2008)
Mann A, et al.