MilliporeSigma
  • Crystal structure of delta-chymotrypsin bound to a peptidyl chloromethyl ketone inhibitor
Crystal structure of delta-chymotrypsin bound to a peptidyl chloromethyl ketone inhibitor
Acta Crystallographica Section D, Biological Crystallography (2000)
Mac Sweeney A, et al.